Learn More
Electron Microscopy Sciences Weibel 2 Graticle NGW2, 16mm
SDP
Catalog No. 5029708
Encompass_Preferred
Used when making a surface to volume ratio of a structure per mass unit. This graticule consists of a number of short lines with interruptions as long as the lines. Basically, the number of intersections falling over the short lines is counted and the number of endpoints falling on the end of the structure is determined. Reference: E.R. Weibel, Journal of Microscopy Vol. 95. pp 373-378. Current Capabilities and Limitations of Available Sterrological Technique, point counting method.
Size: 16 mm
Catalog No. 50-297-08 Supplier Electron Microscopy Sciences Supplier No. 6802516
Used when making a surface to volume ratio of a structure per mass unit. This graticule consists of a number of short lines with interruptions as long as the lines. Basically, the number of intersections falling over the short lines is counted and the number of endpoints falling on the end of the structure is determined. Reference: E.R. Weibel, Journal of Microscopy Vol. 95. pp 373-378. Current Capabilities and Limitations of Available Sterrological Technique, point counting method.
Size: 16 mm