Learn More
Electron Microscopy Sciences Weibel 3 Graticle NGW3, 21mm, Weibel 3 Method
SDP
Catalog No. 5029707
Encompass_Preferred
Weibel Type 3 graticle is used for Stereology. Glass sandwich. Reference: E.R. Weibel, G.S. Kishtler & W.F. Scherle. 1986. J. Cell Biology.30, 23.
Size: 21mm
Method: Weibel 3
Surface: Glass sandwich
Catalog No. 50-297-07 Supplier Electron Microscopy Sciences Supplier No. 6802321
Weibel Type 3 graticle is used for Stereology. Glass sandwich. Reference: E.R. Weibel, G.S. Kishtler & W.F. Scherle. 1986. J. Cell Biology.30, 23.
Size: 21mm
Method: Weibel 3
Surface: Glass sandwich