Learn More
Analytical Sales & Services NH (amino) SPE 96-Well Plate, 2.0mL/25mg Bed
Catalog No. 5000806177
Encompass
NH (amino) SPE, 2mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06177 Supplier Analytical Sales & Services Supplier No. 9620NH25
NH (amino) SPE, 2mL 96 Well Filter Plate, 25mg per well