Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH-bis(C1-Boc) | 85916-13-8 | 245.32 | C12H23NO4 | 500 MG
SDP

Catalog No. 5000202752
Encompass_Preferred

NH-bis(C1-Boc) is an uncleavable Boc-protected linker used in antibody-drug conjugates (ADCs) for research applications. It provides a stable, non-cleavable spacer suitable for conjugation chemistries in ADC development.

  • Used as an uncleavable linker for ADCs.
  • Contains a Boc-protected amine for conjugation reactions.
  • Molecular weight: 245.32 g/mol.
  • Chemical formula: C12H23NO4.
  • CAS number: 85916-13-8.
  • Common pack sizes: 500 mg, 1 g, 5 g, 10 g.

Catalog No. 50-002-02752 Supplier Medchemexpress LLC Supplier No. HY23641500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis(C1-Boc) is an uncleavable Boc-protected linker used in antibody-drug conjugates (ADCs) for research applications. It provides a stable, non-cleavable spacer suitable for conjugation chemistries in ADC development.

  • Used as an uncleavable linker for ADCs.
  • Contains a Boc-protected amine for conjugation reactions.
  • Molecular weight: 245.32 g/mol.
  • Chemical formula: C12H23NO4.
  • CAS number: 85916-13-8.
  • Common pack sizes: 500 mg, 1 g, 5 g, 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.