Learn More
Medchemexpress LLC NH-bis(C2-PEG1-azide) | 2100306-81-6 | C8H17N7O2 | 100 MG
SDP
Catalog No. 5000356900
Encompass_Preferred
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-56900 Supplier Medchemexpress LLC Supplier No. HY140550100MG
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups