Learn More
Medchemexpress LLC NH-bis(C2-PEG1-azide) | 2100306-81-6 | C8H17N7O2 | 250 MG
SDP
Catalog No. 5000352588
Encompass_Preferred
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent with an azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-52588 Supplier Medchemexpress LLC Supplier No. 250MGHY140550
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent with an azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups