Learn More
Medchemexpress LLC NH-bis(C2-PEG1-azide) | 2100306-81-6 | C8H17N7O2 | 50 MG
SDP
Catalog No. 5000357036
Encompass_Preferred
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group that enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-57036 Supplier Medchemexpress LLC Supplier No. HY14055050MG
NH-bis(C2-PEG1-azide) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group that enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions