Learn More
Medchemexpress LLC NH-bis(C2-PEG2-NH-Boc) | 2182601-69-8 | 99.9% | 50 MG
SDP
Catalog No. 5000341418
Encompass_Preferred
NH-bis(C2-PEG2-NH-Boc) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to contain two different ligands connected by a linker, allowing for the exploitation of the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Targets PROTAC linkers and PROTAC pathways.
  • Applicable in cancer-programmed cell death.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-41418 Supplier Medchemexpress LLC Supplier No. 25MGHY130531
NH-bis(C2-PEG2-NH-Boc) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to contain two different ligands connected by a linker, allowing for the exploitation of the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Targets PROTAC linkers and PROTAC pathways.
  • Applicable in cancer-programmed cell death.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.