Learn More
Medchemexpress LLC NH-bis(m-PEG8) | 2055013-64-2 | MFCD28015763 | >98.0% | C34H71NO16 | 250 MG
SDP
Catalog No. 5000352701
Encompass_Preferred
NH-bis(m-PEG8) is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs consist of two ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only
Catalog No. 50-003-52701 Supplier Medchemexpress LLC Supplier No. 250MGHY140264
NH-bis(m-PEG8) is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs consist of two ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only