Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nh-bis-peg2 | 54384-47-3 | MFCD00239212 | 97.0% | 193.24 g·mol⁻¹ | C8H19NO4 | 250 MG
SDP

Catalog No. 5000203016
Encompass_Preferred

NH-bis-PEG2 is a non-cleavable, two-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs). It provides a short, flexible spacer with a terminal amino (NH) functionality suitable for bioconjugation and linker construction.

  • Non-cleavable two-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Provides a short, flexible spacer with terminal NH for bioconjugation.
  • CAS number 54384-47-3.
  • Molecular formula C8H19NO4; molecular weight 193.24 g·mol⁻¹.
  • Purity 97.0%.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Packaged quantity: 250 MG.

Catalog No. 50-002-03016 Supplier Medchemexpress LLC Supplier No. HY130328250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis-PEG2 is a non-cleavable, two-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs). It provides a short, flexible spacer with a terminal amino (NH) functionality suitable for bioconjugation and linker construction.

  • Non-cleavable two-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Provides a short, flexible spacer with terminal NH for bioconjugation.
  • CAS number 54384-47-3.
  • Molecular formula C8H19NO4; molecular weight 193.24 g·mol⁻¹.
  • Purity 97.0%.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Packaged quantity: 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.