Learn More
Medchemexpress LLC Nh-bis(peg2-c2-boc) | 1964503-36-3 | >=98.0% | C22H43NO8 | 25 MG
SDP
Catalog No. 5000352654
Encompass_Preferred
Nh-bis(peg2-c2-boc) is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52654 Supplier Medchemexpress LLC Supplier No. 25MGHY140256
Nh-bis(peg2-c2-boc) is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins