Learn More
Medchemexpress LLC NH-bis(PEG2-propargyl) | 2100306-83-8 | >98% | 269.34 | 250 MG
SDP
Catalog No. 5000348791
Encompass_Preferred
NH-bis(PEG2-propargyl) is a PEG-based PROTAC linker and click chemistry reagent. It has a molecular weight of 269.34 and a purity of >98%.
  • Targets PROTAC linkers
  • Used in PROTAC pathway
  • Soluble in DMSO at 50 mg/mL (ultrasonic)
Catalog No. 50-003-48791 Supplier Medchemexpress LLC Supplier No. 250MGHY140088
Add to Cart
Add to Cart
NH-bis(PEG2-propargyl) is a PEG-based PROTAC linker and click chemistry reagent. It has a molecular weight of 269.34 and a purity of >98%.
  • Targets PROTAC linkers
  • Used in PROTAC pathway
  • Soluble in DMSO at 50 mg/mL (ultrasonic)