Learn More
Medchemexpress LLC NH-bis(PEG3-azide) | 1258939-39-7 | 98.0% | C16H33N7O6 | 50 MG
SDP
Catalog No. 5000356664
Encompass_Preferred
NH-bis(PEG3-azide) is a PEG-based PROTAC linker used in PROTAC synthesis. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Can be used in the synthesis of PROTACs.
  • Contains an azide group, facilitating click chemistry reactions.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-56664 Supplier Medchemexpress LLC Supplier No. HY14055150MG
NH-bis(PEG3-azide) is a PEG-based PROTAC linker used in PROTAC synthesis. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Can be used in the synthesis of PROTACs.
  • Contains an azide group, facilitating click chemistry reactions.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.