Learn More
Medchemexpress LLC NH-bis(PEG3-Boc) | 1814901-03-5 | 98.0% | C26H51NO10 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1402571G
Encompass_Preferred
NH-bis(PEG3-Boc) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds designed with two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins. This product appears as a colorless to light yellow liquid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Colorless to light yellow liquid
Catalog No. 50-003-53510