Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nh-bis(peg4-c2-nh-boc) | 2182601-75-6 | ≥97.0% | C30H61N3O12 | 500 MG
SDP

Catalog No. 5000357235
Encompass_Preferred

NH-bis(PEG4-C2-NH-Boc) is a PEG-based linker utilized in PROTAC synthesis, appearing as a colorless to light yellow liquid. This compound is intended for research use only and has not been fully validated for medical applications.

  • PEG-based linker for PROTAC synthesis.
  • Purity of at least 97.0% as determined by NMR.
  • Stable when stored at -20°C for up to 3 years or 4°C for up to 2 years in pure form.
  • Maintain stability for up to 6 months at -80°C or 1 month at -20°C when in solvent.

Catalog No. 50-003-57235 Supplier Medchemexpress LLC Supplier No. HY140261500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis(PEG4-C2-NH-Boc) is a PEG-based linker utilized in PROTAC synthesis, appearing as a colorless to light yellow liquid. This compound is intended for research use only and has not been fully validated for medical applications.

  • PEG-based linker for PROTAC synthesis.
  • Purity of at least 97.0% as determined by NMR.
  • Stable when stored at -20°C for up to 3 years or 4°C for up to 2 years in pure form.
  • Maintain stability for up to 6 months at -80°C or 1 month at -20°C when in solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.