Learn More
Medchemexpress LLC HY-100576 100mg Medchemexpress, NH125 CAS:278603-08-0 Purity:>98%
SDP
Catalog No. 501931889
Encompass_Preferred
Medchemexpress, HY-100576 100mg NH125 CAS:278603-08-0 NH125 is a potent and selective inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII) , also could induce eEF2 phosphorylation, with an IC 50 of 60 nM for eEF-2K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1889 Supplier Medchemexpress LLC Supplier No. HY100576100MG
Medchemexpress, HY-100576 100mg NH125 CAS:278603-08-0 NH125 is a potent and selective inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII) , also could induce eEF2 phosphorylation, with an IC 50 of 60 nM for eEF-2K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.