Learn More
Medchemexpress LLC Nh125 | 278603-08-0 | MFCD07370143 | >=98.0% | 524.56 g/mol | C27H45IN2 | 25 MG
SDP
Catalog No. 5000212106
Encompass_Preferred
NH125 is a small-molecule inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII) for laboratory research. It inhibits eEF-2K with an IC50 of 60 nM and blocks eEF2 phosphorylation in intact cells, enabling studies of protein synthesis regulation, autophagy, and kinase signaling.
  • Potent inhibition of eEF-2K (IC50 ≈ 60 nM) for biochemical assays.
  • Cell-active: blocks eEF2 phosphorylation in intact cells.
  • Suitable for studies of translation regulation, autophagy, and kinase signaling.
  • Available in solid and DMSO solution formats for assay flexibility.
  • Storage recommendations support short-term and long-term stability when handled correctly.
Catalog No. 50-002-12106 Supplier Medchemexpress LLC Supplier No. HY10057625MG
NH125 is a small-molecule inhibitor of eukaryotic elongation factor 2 kinase (eEF-2K/CaMKIII) for laboratory research. It inhibits eEF-2K with an IC50 of 60 nM and blocks eEF2 phosphorylation in intact cells, enabling studies of protein synthesis regulation, autophagy, and kinase signaling.
  • Potent inhibition of eEF-2K (IC50 ≈ 60 nM) for biochemical assays.
  • Cell-active: blocks eEF2 phosphorylation in intact cells.
  • Suitable for studies of translation regulation, autophagy, and kinase signaling.
  • Available in solid and DMSO solution formats for assay flexibility.
  • Storage recommendations support short-term and long-term stability when handled correctly.