Learn More
YMC America, Inc NH2 120A 3um 150x0.15mm 1/16
SDP
Catalog No. 5000881272
Encompass
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-81272 Supplier YMC America, Inc Supplier No. NH12S0315F5AU
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange