Learn More
YMC America, Inc NH2 120A 5um 75x0.3mm 1/32
SDP
Catalog No. 5000877156
Encompass
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-77156 Supplier YMC America, Inc Supplier No. NH12S05L5H0RU
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange