Learn More
YMC America, Inc NH2 300A 5um 100x0.15mm 1/16
SDP
Catalog No. 5000880841
Encompass
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-80841 Supplier YMC America, Inc Supplier No. NH30S0510F5AU
Add to Cart
Add to Cart
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange