Learn More
Medchemexpress LLC NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT | 2857037-69-3 | 98.7% | 473.48 | 25 MG
SDP
Catalog No. 5000363576
Encompass_Preferred
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT (compound I) is a topoisomerase I inhibitor, which can be delivered to cells through conjugated antibody targeting, and has good ADC activity in vivo and in vitro. It is for research use only and not sold to patients.
  • Topoisomerase I inhibitor
  • Delivers through conjugated antibody targeting
  • Good ADC activity in vivo and in vitro
  • For research use only
Catalog No. 50-003-63576 Supplier Medchemexpress LLC Supplier No. 25MGHY148819
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT (compound I) is a topoisomerase I inhibitor, which can be delivered to cells through conjugated antibody targeting, and has good ADC activity in vivo and in vitro. It is for research use only and not sold to patients.
  • Topoisomerase I inhibitor
  • Delivers through conjugated antibody targeting
  • Good ADC activity in vivo and in vitro
  • For research use only