Learn More
Medchemexpress LLC NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT | 2857037-69-3 | 98.7% | 473.48 | 5 MG
SDP
Catalog No. 5000360844
Encompass_Preferred
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT is a potent topoisomerase I inhibitor. It is designed for delivery to cells via conjugated antibody targeting, demonstrating strong antibody-drug conjugate (ADC) activity both in vivo and in vitro.
  • Functions as a topoisomerase I inhibitor
  • Designed for conjugated antibody targeting
  • Exhibits good ADC activity in vivo
  • Shows good ADC activity in vitro
Catalog No. 50-003-60844 Supplier Medchemexpress LLC Supplier No. HY1488195MG
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT is a potent topoisomerase I inhibitor. It is designed for delivery to cells via conjugated antibody targeting, demonstrating strong antibody-drug conjugate (ADC) activity both in vivo and in vitro.
  • Functions as a topoisomerase I inhibitor
  • Designed for conjugated antibody targeting
  • Exhibits good ADC activity in vivo
  • Shows good ADC activity in vitro