Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT | 2857037-69-3 | 98.7% | 473.48 g·mol⁻¹ | C26H23N3O6 | 10 MG
SDP

Catalog No. 5000285383
Encompass_Preferred

NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT is a small-molecule topoisomerase I inhibitor used in research as an antibody-drug conjugate (ADC) payload; the compound shows demonstrated activity in vitro and in vivo and is supplied for laboratory studies.

  • High purity (98.7%) suitable for research applications.
  • Solid form, provided as a dry material for formulation or conjugation.
  • Molecular formula C26H23N3O6; molecular weight 473.48 g·mol⁻¹.
  • Available in 10 mg pack sizes for laboratory-scale experiments.
  • Store at -20°C and protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-85383 Supplier Medchemexpress LLC Supplier No. HY14881910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT is a small-molecule topoisomerase I inhibitor used in research as an antibody-drug conjugate (ADC) payload; the compound shows demonstrated activity in vitro and in vivo and is supplied for laboratory studies.

  • High purity (98.7%) suitable for research applications.
  • Solid form, provided as a dry material for formulation or conjugation.
  • Molecular formula C26H23N3O6; molecular weight 473.48 g·mol⁻¹.
  • Available in 10 mg pack sizes for laboratory-scale experiments.
  • Store at -20°C and protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.