Learn More
Medchemexpress LLC NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT hydrochloride | 98.5% | 509.94 | 25 MG
SDP
Catalog No. 5000363577
Encompass_Preferred
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT (compound I) hydrochloride is a topoisomerase I inhibitor that demonstrates good ADC (Antibody-Drug Conjugate) activity both in vivo and in vitro, and can be delivered to cells through conjugated antibody targeting.
  • Functions as a topoisomerase I inhibitor.
  • Designed for targeted delivery to cells via conjugated antibodies.
  • Shows effective ADC activity both in vivo and in vitro.
Catalog No. 50-003-63577 Supplier Medchemexpress LLC Supplier No. 25MGHY148819A
NH2-bicyclo[1.1.1]pentane-7-MAD-MDCPT (compound I) hydrochloride is a topoisomerase I inhibitor that demonstrates good ADC (Antibody-Drug Conjugate) activity both in vivo and in vitro, and can be delivered to cells through conjugated antibody targeting.
  • Functions as a topoisomerase I inhibitor.
  • Designed for targeted delivery to cells via conjugated antibodies.
  • Shows effective ADC activity both in vivo and in vitro.