Learn More
Medchemexpress LLC NH2-C2-NH-Boc (PROTAC Linker 22) | 57260-73-8 | 99.97% | 160.21 | 10 G
SDP
Catalog No. 5000404806
Encompass_Preferred
NH2-C2-NH-Boc (PROTAC Linker 22) is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Can be used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-04806 Supplier Medchemexpress LLC Supplier No. HY4017110G
NH2-C2-NH-Boc (PROTAC Linker 22) is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Can be used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins