Learn More
Medchemexpress LLC NH2-C5-NH-Boc | 51644-96-3 | 202.29 | 10 G
SDP
Catalog No. 5000449070
Encompass_Preferred
NH2-C5-NH-Boc (PROTAC Linker 23) is an alkyl chain-based PROTAC linker. It is used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-49070 Supplier Medchemexpress LLC Supplier No. HYW00471010G
NH2-C5-NH-Boc (PROTAC Linker 23) is an alkyl chain-based PROTAC linker. It is used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins