Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-C5-PEG4-N3-L-lysine-PEG3-N3 | 99.4% | 619.71 | C25H49N9O9 | 50 MG
SDP

Catalog No. 5000204037
Encompass_Preferred

NH2-C5-PEG4-N3-L-lysine-PEG3-N3 is a cleavable 7-unit PEG antibody-drug conjugate (ADC) linker containing an azide functional group, intended for research use in click chemistry conjugations. It facilitates attachment of payloads to biomolecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC).

  • Cleavable 7-unit PEG linker suitable for ADC synthesis.
  • Azide functional group for CuAAC and SPAAC click reactions.
  • High purity (≈99.4%).
  • Water soluble: ≥100 mg/mL.
  • Appearance: colorless to light yellow liquid.
  • Recommended storage: pure form at -20°C.
  • Packaged in 50 mg units.

Catalog No. 50-002-04037 Supplier Medchemexpress LLC Supplier No. HY13094650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-C5-PEG4-N3-L-lysine-PEG3-N3 is a cleavable 7-unit PEG antibody-drug conjugate (ADC) linker containing an azide functional group, intended for research use in click chemistry conjugations. It facilitates attachment of payloads to biomolecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC).

  • Cleavable 7-unit PEG linker suitable for ADC synthesis.
  • Azide functional group for CuAAC and SPAAC click reactions.
  • High purity (≈99.4%).
  • Water soluble: ≥100 mg/mL.
  • Appearance: colorless to light yellow liquid.
  • Recommended storage: pure form at -20°C.
  • Packaged in 50 mg units.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.