Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-C5-PEG4-N3-L-lysine-PEG3-N3 | 99.4% | 619.71 g/mol | C25H49N9O9 | 100 MG
SDP

Catalog No. 5000202177
Encompass_Preferred

A cleavable 7-unit polyethylene glycol (PEG) linker with an amino terminus and azide functionality used for antibody-drug conjugate (ADC) synthesis and bioconjugation workflows.

  • Cleavable 7-unit PEG linker suitable for ADC synthesis.
  • Contains amino (NH2) and azide (N3) functional groups for conjugation.
  • High purity: 99.4%.
  • Molecular weight: 619.71 g/mol.
  • Soluble ≥ 100 mg/mL in water for aqueous conjugations.
  • Available in multiple pack sizes, including 100 MG.

Catalog No. 50-002-02177 Supplier Medchemexpress LLC Supplier No. HY130946100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cleavable 7-unit polyethylene glycol (PEG) linker with an amino terminus and azide functionality used for antibody-drug conjugate (ADC) synthesis and bioconjugation workflows.

  • Cleavable 7-unit PEG linker suitable for ADC synthesis.
  • Contains amino (NH2) and azide (N3) functional groups for conjugation.
  • High purity: 99.4%.
  • Molecular weight: 619.71 g/mol.
  • Soluble ≥ 100 mg/mL in water for aqueous conjugations.
  • Available in multiple pack sizes, including 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.