Learn More
Medchemexpress LLC NH2-C6-ARC186 sodium | 5 MG
SDP
Catalog No. 5000376301
Encompass_Preferred
NH2-C6-ARC186 sodium is a modified ARC186 with NH2-C6, enabling coupling to other peptides or molecules. It functions as a potent complement inhibitor, specifically an aptamer, by blocking the convertase-catalyzed activation of C5.
  • Modified ARC186 with NH2-C6
  • Can be coupled to other peptides or molecules
  • Potent complement inhibitor
  • Aptamer that blocks convertase-catalyzed activation of C5
  • Targets the Complement System
  • Relevant to Immunology/Inflammation
  • Store at -20°C in sealed storage, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture
Catalog No. 50-003-76301 Supplier Medchemexpress LLC Supplier No. HY1537855MG
NH2-C6-ARC186 sodium is a modified ARC186 with NH2-C6, enabling coupling to other peptides or molecules. It functions as a potent complement inhibitor, specifically an aptamer, by blocking the convertase-catalyzed activation of C5.
  • Modified ARC186 with NH2-C6
  • Can be coupled to other peptides or molecules
  • Potent complement inhibitor
  • Aptamer that blocks convertase-catalyzed activation of C5
  • Targets the Complement System
  • Relevant to Immunology/Inflammation
  • Store at -20°C in sealed storage, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture