Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-C6-ARC186 sodium | 92.5% | 12,881.00 | 10 MG
SDP

Catalog No. 5000287094
Encompass_Preferred

NH2-C6-ARC186 sodium is an NH2-C6-modified aptamer supplied as the sodium salt that inhibits complement C5 convertase activity. The terminal aminohexyl (NH2-C6) linker enables covalent coupling to other peptides or molecules for conjugation studies. It is provided as a white to off-white solid with defined purity and molecular weight for research applications in small milligram quantities.

  • Function: complement C5 convertase inhibitor
  • NH2-C6 aminohexyl linker for conjugation
  • Purity: 92.5%
  • Physical form: white to off-white solid
  • Typical sizes: 1 mg, 5 mg, 10 mg, larger sizes by quote
  • Storage: in solvent -80°C (6 months) or -20°C (1 month); sealed and dry
  • Molecular weight approximately 12,881 Da

Catalog No. 50-002-87094 Supplier Medchemexpress LLC Supplier No. HY15378510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-C6-ARC186 sodium is an NH2-C6-modified aptamer supplied as the sodium salt that inhibits complement C5 convertase activity. The terminal aminohexyl (NH2-C6) linker enables covalent coupling to other peptides or molecules for conjugation studies. It is provided as a white to off-white solid with defined purity and molecular weight for research applications in small milligram quantities.

  • Function: complement C5 convertase inhibitor
  • NH2-C6 aminohexyl linker for conjugation
  • Purity: 92.5%
  • Physical form: white to off-white solid
  • Typical sizes: 1 mg, 5 mg, 10 mg, larger sizes by quote
  • Storage: in solvent -80°C (6 months) or -20°C (1 month); sealed and dry
  • Molecular weight approximately 12,881 Da

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.