Learn More
Medchemexpress LLC NH2-C6-NH-Boc | 51857-17-1 | 216.32 | 500 G
SDP
Catalog No. 5000449396
Encompass_Preferred
NH2-C6-NH-Boc is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of Mcl-1 inhibitors based on PROTAC technology. This product is for research use only.
  • PROTAC linker
  • Alkyl/ether composition
  • Used in synthesis of Mcl-1 inhibitors
Catalog No. 50-004-49396 Supplier Medchemexpress LLC Supplier No. HYW008296500G
NH2-C6-NH-Boc is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of Mcl-1 inhibitors based on PROTAC technology. This product is for research use only.
  • PROTAC linker
  • Alkyl/ether composition
  • Used in synthesis of Mcl-1 inhibitors