Learn More
YMC America, Inc NH2 col 120A 5um 35x4.6mm
SDP
Catalog No. 5000880827
Encompass_Preferred
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-80827 Supplier YMC America, Inc Supplier No. NH12S05H546WT
Add to Cart
Add to Cart
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange