Learn More
Medchemexpress LLC NH2 DOTA-GA | 1639843-65-4 | 518.56 | C21H38N6O9 | 50 MG
SDP
Catalog No. 5000202435
Encompass_Preferred
NH2 DOTA-GA is a DOTA-based bifunctional metal chelator used to prepare radionuclide drug conjugates (RDCs), with carboxylic acid groups capable of complexing metal ions such as Gd(III).
  • Bifunctional DOTA chelator for radiolabeling of biomolecules.
  • Suitable for preparation of radionuclide-drug conjugates.
  • Molecular formula C21H38N6O9; molecular weight 518.56.
  • Available as a 50 mg powder; stable for 3 years at -20°C when stored as recommended.
  • Listed CAS number 1639843-65-4 for unambiguous identification.
Catalog No. 50-002-02435 Supplier Medchemexpress LLC Supplier No. HY16456950MG
NH2 DOTA-GA is a DOTA-based bifunctional metal chelator used to prepare radionuclide drug conjugates (RDCs), with carboxylic acid groups capable of complexing metal ions such as Gd(III).
  • Bifunctional DOTA chelator for radiolabeling of biomolecules.
  • Suitable for preparation of radionuclide-drug conjugates.
  • Molecular formula C21H38N6O9; molecular weight 518.56.
  • Available as a 50 mg powder; stable for 3 years at -20°C when stored as recommended.
  • Listed CAS number 1639843-65-4 for unambiguous identification.