Learn More
Medchemexpress LLC Nh2 DOTA-GA | 1639843-65-4 | 518.56 g/mol | C21H38N6O9 | 100 MG
SDP
Catalog No. 5000201937
Encompass_Preferred
NH2 DOTA-GA is an amino-functionalized DOTA-based metal chelator used to prepare radionuclide drug conjugates (RDCs). Its carboxylic acid groups complex metal ions (for example, Gd(III)), enabling radiolabeling and imaging agent development.
  • Amino-functionalized chelator suitable for conjugation.
  • Forms stable complexes with metal ions such as Gd(III).
  • Used in synthesis of radionuclide drug conjugates.
  • Available in multiple research-scale sizes, including 100 mg.
  • Molecular weight 518.56 g/mol and formula C21H38N6O9.
Catalog No. 50-002-01937 Supplier Medchemexpress LLC Supplier No. HY164569100MG
NH2 DOTA-GA is an amino-functionalized DOTA-based metal chelator used to prepare radionuclide drug conjugates (RDCs). Its carboxylic acid groups complex metal ions (for example, Gd(III)), enabling radiolabeling and imaging agent development.
  • Amino-functionalized chelator suitable for conjugation.
  • Forms stable complexes with metal ions such as Gd(III).
  • Used in synthesis of radionuclide drug conjugates.
  • Available in multiple research-scale sizes, including 100 mg.
  • Molecular weight 518.56 g/mol and formula C21H38N6O9.