Learn More
YMC America, Inc NH2-HG col 120A 10um 125x4mm
SDP
Catalog No. 5000880277
Encompass_Preferred
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-80277 Supplier YMC America, Inc Supplier No. NHG12S11R504WT
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange