Learn More
YMC America, Inc NH2-HG guard 120A 10um 10x4mm
SDP
Catalog No. 5000881222
Encompass
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange
Catalog No. 50-008-81222 Supplier YMC America, Inc Supplier No. NHG12S110104GC
YMC-Pack NH2 is a normal phase separation column utilizing the polarity of primary amino groups bonded to silica It is a versatile stationary phase often used for the separation of sugars but can also be used for separations utilizing weak anion-exchange