Learn More
Medchemexpress LLC HY-W067489 100mg Medchemexpress, NH2-PEG1-CH2CH2-Boc CAS:1260092-46-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW067489100MG
Encompass_Preferred
Medchemexpress, HY-W067489 100mg NH2-PEG1-CH2CH2-Boc CAS:1260092-46-3 NH2-PEG1-CH2CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3499