Learn More
Medchemexpress LLC HY-128801 100mg Medchemexpress, NH2-PEG3-C1-Boc CAS:189808-70-6 Purity:>98%
SDP
Catalog No. 502081185
Encompass_Preferred
Medchemexpress, HY-128801 100mg NH2-PEG3-C1-Boc CAS:189808-70-6 NH2-PEG3-C1-Boc (PROTAC Linker 5) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1185 Supplier Medchemexpress LLC Supplier No. HY128801100MG
Medchemexpress, HY-128801 100mg NH2-PEG3-C1-Boc CAS:189808-70-6 NH2-PEG3-C1-Boc (PROTAC Linker 5) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.