Learn More
Medchemexpress LLC NH2-PEG3-C6-Cl | 1261350-60-0 | 267.79 | 50 MG
SDP
Catalog No. 5000350296
Encompass_Preferred
NH2-PEG3-C6-Cl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • One is a ligand for an E3 ubiquitin ligase
  • The other is for the target protein
Catalog No. 50-003-50296 Supplier Medchemexpress LLC Supplier No. HY13846950MG
NH2-PEG3-C6-Cl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • One is a ligand for an E3 ubiquitin ligase
  • The other is for the target protein