Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-PEG3 (PROTAC Linker 35) | 6338-55-2 | 149.19 | 25 G
SDP

Catalog No. 5000449850
Encompass_Preferred

NH2-PEG3 (PROTAC Linker 35) is a polyethylene glycol (PEG) linker used in the synthesis of PROTACs such as β-NF-JQ1. This chemical is intended for research use only.

  • PROTAC linker
  • Polyethylene glycol linker
  • Used in the synthesis of PROTAC (β-NF-JQ1)
  • Purity: 99.9%
  • Molecular weight: 149.19
  • Formula: C6H15NO3
  • Appearance: Liquid (density: 1.0773 g/cm3)
  • Color: Colorless to light yellow

Catalog No. 50-004-49850 Supplier Medchemexpress LLC Supplier No. 25GHYW007545
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-PEG3 (PROTAC Linker 35) is a polyethylene glycol (PEG) linker used in the synthesis of PROTACs such as β-NF-JQ1. This chemical is intended for research use only.

  • PROTAC linker
  • Polyethylene glycol linker
  • Used in the synthesis of PROTAC (β-NF-JQ1)
  • Purity: 99.9%
  • Molecular weight: 149.19
  • Formula: C6H15NO3
  • Appearance: Liquid (density: 1.0773 g/cm3)
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.