Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nh2-peg3 (protac linker 35) | 6338-55-2 | MFCD07367495 | 149.19 | 1 G
SDP

Catalog No. 5000446588
Encompass_Preferred

NH2-PEG3 (PROTAC Linker 35) is a PROTAC linker, which belongs to a polyethylene glycol (PEG) linker. This compound can be used in the synthesis of the PROTAC (β-NF-JQ1).

  • PROTAC linker
  • Polyethylene glycol (PEG) linker
  • Suitable for synthesis of PROTAC (β-NF-JQ1)
  • Colorless to light yellow liquid
  • Purity: 99.87%
  • Molecular weight: 149.19
  • CAS number: 6338-55-2
  • Density: 1.0773 g/cm3
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-46588 Supplier Medchemexpress LLC Supplier No. HYW0075451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-PEG3 (PROTAC Linker 35) is a PROTAC linker, which belongs to a polyethylene glycol (PEG) linker. This compound can be used in the synthesis of the PROTAC (β-NF-JQ1).

  • PROTAC linker
  • Polyethylene glycol (PEG) linker
  • Suitable for synthesis of PROTAC (β-NF-JQ1)
  • Colorless to light yellow liquid
  • Purity: 99.87%
  • Molecular weight: 149.19
  • CAS number: 6338-55-2
  • Density: 1.0773 g/cm3
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.