Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NH2-PEG4-CH2CH2COOH | 663921-15-1 | 99.9% | 265.30 g/mol | C11H23NO6 | 500 MG
SDP

Catalog No. 5000202846
Encompass_Preferred

NH2-PEG4-CH2CH2COOH is a cleavable four-unit polyethylene glycol (PEG4) linker with terminal amino and carboxylic acid functional groups used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. The material is supplied as a solid with high purity and shows good solubility in DMSO.

  • Cleavable PEG4 linker with terminal amino and carboxyl groups
  • Purity 99.9%
  • CAS number 663921-15-1
  • Molecular weight 265.30 g/mol
  • Solubility: DMSO (10 mg/mL with warming)
  • Pack size: 500 MG

Catalog No. 50-002-02846 Supplier Medchemexpress LLC Supplier No. HYW021787500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH2-PEG4-CH2CH2COOH is a cleavable four-unit polyethylene glycol (PEG4) linker with terminal amino and carboxylic acid functional groups used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. The material is supplied as a solid with high purity and shows good solubility in DMSO.

  • Cleavable PEG4 linker with terminal amino and carboxyl groups
  • Purity 99.9%
  • CAS number 663921-15-1
  • Molecular weight 265.30 g/mol
  • Solubility: DMSO (10 mg/mL with warming)
  • Pack size: 500 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.