Learn More
Medchemexpress LLC Nh2-peg4-dota | 2090232-34-9 | 97.0% | 622.71 | C26H50N6O11 | 25mg
SDP
Catalog No. 5000270413
Encompass_Preferred
NH2-PEG4-DOTA is a bifunctional DOTA-type metal chelator NH2-PEG4-DOTA can be conjugated with Dextran for quantitative analysis in the presence of a highly luminescent complex NH2-PEG4-DOTA can also bind to radionuclides to prepare radionuclide drug conjugates (RDCs) RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy
Catalog No. 50-002-70413 Supplier Medchemexpress LLC Supplier No. HYW75503325MG
NH2-PEG4-DOTA is a bifunctional DOTA-type metal chelator NH2-PEG4-DOTA can be conjugated with Dextran for quantitative analysis in the presence of a highly luminescent complex NH2-PEG4-DOTA can also bind to radionuclides to prepare radionuclide drug conjugates (RDCs) RDCs have the ability to specifically target biomolecules and can be used in medical imaging or therapy