Learn More
Medchemexpress LLC NH2-PEG5-C6-Cl hydrochloride | 2241669-16-7 | 98.0% | 1 G
SDP
Catalog No. 5000338431
Encompass_Preferred
NH2-PEG5-C6-Cl hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Applicable in cancer and cancer targeted therapy research
  • Classified as PROTAC linkers
Catalog No. 50-003-38431 Supplier Medchemexpress LLC Supplier No. HY129622A1G
NH2-PEG5-C6-Cl hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Applicable in cancer and cancer targeted therapy research
  • Classified as PROTAC linkers