Learn More
Medchemexpress LLC Nh2-peg5-c6-cl hydrochloride | 2241669-16-7 | 98.0% | 5 G
SDP
Catalog No. 5000339714
Encompass_Preferred
NH2-PEG5-C6-Cl hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-39714 Supplier Medchemexpress LLC Supplier No. HY129622A5G
NH2-PEG5-C6-Cl hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins