Learn More
Medchemexpress LLC NH2-PEG6-Boc | 1286281-32-0 | 98.0% | 25 G
SDP
Catalog No. 5000341778
Encompass_Preferred
NH2-PEG6-Boc is a PEG-based linker used in the synthesis of PROTACs and antibody-drug conjugates (ADCs). It functions as a non-cleavable 6-unit PEG ADC linker, facilitating the degradation of target proteins via the ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Non-cleavable 6 unit PEG ADC linker
  • Used in the synthesis of PROTACs
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41778 Supplier Medchemexpress LLC Supplier No. 25GHY130486
NH2-PEG6-Boc is a PEG-based linker used in the synthesis of PROTACs and antibody-drug conjugates (ADCs). It functions as a non-cleavable 6-unit PEG ADC linker, facilitating the degradation of target proteins via the ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Non-cleavable 6 unit PEG ADC linker
  • Used in the synthesis of PROTACs
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins