Learn More
Medchemexpress LLC Nh2-PEG6-CH2CH2COOH | 905954-28-1 | 353.41 | C15H31NO8 | 250 MG
SDP
Catalog No. 5000203214
Encompass_Preferred
NH2-PEG6-CH2CH2COOH is a cleavable six-unit polyethylene glycol (PEG) linker with an amino terminus and a terminal carboxyethyl group, intended for use in antibody-drug conjugate linker chemistry, PROTAC linker synthesis, and other bioconjugation applications.
  • Cleavable six-unit PEG spacer provides hydrophilic separation between payload and targeting moiety.
  • Amino terminus and carboxylic acid group enable diverse conjugation chemistries.
  • Suitable for ADC and PROTAC linker development and general bioconjugation workflows.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-03214 Supplier Medchemexpress LLC Supplier No. HYW040257250MG
NH2-PEG6-CH2CH2COOH is a cleavable six-unit polyethylene glycol (PEG) linker with an amino terminus and a terminal carboxyethyl group, intended for use in antibody-drug conjugate linker chemistry, PROTAC linker synthesis, and other bioconjugation applications.
  • Cleavable six-unit PEG spacer provides hydrophilic separation between payload and targeting moiety.
  • Amino terminus and carboxylic acid group enable diverse conjugation chemistries.
  • Suitable for ADC and PROTAC linker development and general bioconjugation workflows.
  • Available in small research pack sizes for laboratory use.