Learn More
Medchemexpress LLC HY-120918 250mg Medchemexpress, NH2-PEG7 CAS:1425973-14-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120918250MG
Encompass_Preferred
Medchemexpress, HY-120918 250mg NH2-PEG7 CAS:1425973-14-3 NH2-PEG7 is a PROTAC linker, which refers to the PEG composition. NH2-PEG7 can be used in the synthesis of the PROTAC PARP1 degrader iRucaparib-AP6. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0958