Learn More
Medchemexpress LLC NH2-PEG8-acid | 100mg
SDP
Catalog No. 5000203953
Encompass_Preferred
NH2-PEG8-acid is a non-cleavable 8 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1] NH2-PEG8-acid also is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[2]
Catalog No. 50-002-03953 Supplier Medchemexpress LLC Supplier No. HYW019798100MG
NH2-PEG8-acid is a non-cleavable 8 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1] NH2-PEG8-acid also is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[2]