Learn More
Medchemexpress LLC NH2-PEG8-OH | 352439-37-3 | 97.0% | 100 MG
SDP
Catalog No. 5000339646
Encompass_Preferred
NH2-PEG8-OH is a laboratory chemical used for the manufacture of various substances. This compound is a PEG-based PROTAC linker with a molecular formula of C16H35NO8 and a molecular weight of 369.45. It appears as a colorless to off-white solid-liquid mixture and is stable under recommended storage conditions.
  • High purity (97.0% by NMR)
  • Structural consistency confirmed by 1H NMR and MS
  • Recommended storage at 4°C, protected from light
  • Suitable for research applications
Catalog No. 50-003-39646 Supplier Medchemexpress LLC Supplier No. HY130200100MG
NH2-PEG8-OH is a laboratory chemical used for the manufacture of various substances. This compound is a PEG-based PROTAC linker with a molecular formula of C16H35NO8 and a molecular weight of 369.45. It appears as a colorless to off-white solid-liquid mixture and is stable under recommended storage conditions.
  • High purity (97.0% by NMR)
  • Structural consistency confirmed by 1H NMR and MS
  • Recommended storage at 4°C, protected from light
  • Suitable for research applications